siRNA Details
| Gene Symbol: FAM173A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271285.1 | NP_001258214.1 | 2 | 334 | ||
| NM_023933.2 | NP_076422.1 | 2 | 334 | |||
| GenBank mRNA | AK123186.1 | - | 1 | 1056 | ||
| BC001181.1 | - | 2 | 318 | 3355322 | ||
| BC002624.2 | - | 2 | 286 | 3606040 | ||
| BI597021.1 | - | 2 | 339 | |||
| BI838160.1 | - | 2 | 317 | |||

FAM173A
Genomic Map