siRNA Details
| Gene Symbol: SMARCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003076.4 | NP_003067.3 | 10 | 1416 | ||
| NM_139071.2 | NP_620710.2 | 10 | 1416 | |||
| XM_005269107.3 | XP_005269164.2 | 9 | 1293 | |||
| GenBank mRNA | AF109733.1 | - | 10 | 1204 | ||
| AK130517.1 | - | 11 | 1156 | |||
| AK292700.1 | - | 10 | 1080 | |||
| BC009368.2 | - | 10 | 1366 | 4110499 | ||
| N75126.1 | - | 1 | 126 | |||
| U66617.1 | - | 10 | 1394 | |||

SMARCD1
Genomic Map