siRNA Details
Gene Symbol: SNRPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003095.2 | NP_003086.1 | 2, 3 | 270 | ||
GenBank mRNA | AK311752.1 | - | 2, 3 | 233 | ||
BC002505.2 | - | 2, 3 | 263 | 3051263 | ||
BC063397.1 | - | 2, 3 | 220 | 5176590 | ||
BC128452.1 | - | 2, 3 | 173 | 40123237 | ||
BC128453.1 | - | 2, 3 | 172 | 40123239 | ||
BM972965.1 | - | 2, 3 | 533 | |||
BU861031.1 | - | 2, 3 | 273 | |||
CR456751.1 | - | 1, 2 | 127 | |||
X85372.1 | - | 2, 3 | 242 |
