siRNA Details
| Gene Symbol: SNTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003098.2 | NP_003089.1 | 4 | 1142 | ||
| XM_005260517.1 | XP_005260574.1 | 4 | 1103 | |||
| XM_011529007.1 | XP_011527309.1 | 4 | 1103 | |||
| XM_011529008.1 | XP_011527310.1 | 4 | 1103 | |||
| GenBank mRNA | AK095942.1 | - | 4 | 702 | ||
| AK291994.1 | - | 4 | 1013 | |||
| BC026215.2 | - | 4 | 982 | 4413197 | ||
| BC113813.1 | - | 2 | 290 | |||
| S81737.1 | - | 4 | 1002 | |||
| U40571.1 | - | 4 | 907 | |||

SNTA1
Genomic Map