siRNA Details
| Gene Symbol: SP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017371.4 | NP_001017371.3 | 2 | 1362 | ||
| NM_001172712.1 | NP_001166183.1 | 4 | 1704 | |||
| NM_003111.4 | NP_003102.1 | 4 | 1713 | |||
| GenBank mRNA | AB209334.1 | - | 1 | 826 | ||
| AK304199.1 | - | 4 | 1285 | |||
| AY070137.1 | - | 2 | 1362 | |||
| AY441957.1 | - | 4 | 1713 | |||
| AY441958.1 | - | 2 | 1052 | |||
| BC126414.1 | - | 4 | 1235 | 8992130 | ||
| BC143928.1 | - | 4 | 1226 | 9052446 | ||
| M97191.2 | - | 2 | 973 | |||
| X68560.1 | - | 2 | 931 | |||

SP3
Genomic Map