siRNA Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001309434.1 | NP_001296363.1 | 2 | 164 | ||
| NM_003134.5 | NP_003125.3 | 2 | 164 | |||
| GenBank mRNA | AB061546.1 | - | 2 | 151 | ||
| AB451391.1 | - | 2 | 78 | |||
| BC035495.2 | - | 2 | 120 | 4298087 | ||
| BC071716.1 | - | 2 | 100 | 4476669 | ||
| BC100031.1 | - | 2 | 104 | 2847600 | ||
| DA338824.1 | - | 2 | 164 | |||
| U07857.1 | - | 2 | 99 | |||
| X73459.1 | - | 2 | 97 | |||

SRP14
Genomic Map