siRNA Details
| Gene Symbol: SRPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278273.1 | NP_001265202.1 | 16 | 1994 | ||
| NM_182691.2 | NP_872633.1 | 14 | 2047 | |||
| NM_182692.2 | NP_872634.1 | 15 | 1970 | |||
| XM_005250549.3 | XP_005250606.2 | 16 | 2229 | |||
| XM_005250550.3 | XP_005250607.1 | 15 | 1974 | |||
| XM_005250551.4 | XP_005250608.1 | 16 | 2022 | |||
| XM_006716098.2 | XP_006716161.1 | 16 | 1989 | |||
| XM_011516536.2 | XP_011514838.1 | 14 | 1985 | |||
| XM_011516538.2 | XP_011514840.1 | 15 | 1858 | |||
| XM_017012560.1 | XP_016868049.1 | 16 | 2044 | |||
| XM_017012562.1 | XP_016868051.1 | 16 | 2228 | |||
| XM_017012564.1 | XP_016868053.1 | 14 | 2033 | |||
| XM_017012566.1 | XP_016868055.1 | 16 | 2030 | |||
| GenBank mRNA | AK093420.1 | - | 6 | 1769 | ||
| AY354201.1 | - | 15 | 2136 | |||
| BC035214.1 | - | 14 | 1966 | 4778251 | ||
| BC068547.1 | - | 16 | 1994 | 5265599 | ||
| U88666.1 | - | 14 | 1997 | |||

SRPK2
Genomic Map