siRNA Details
| Gene Symbol: SSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308197.1 | NP_001295126.1 | 5 | 658 | ||
| NM_001308204.1 | NP_001295133.1 | 5 | 484 | |||
| NM_001308205.1 | NP_001295134.1 | 5 | 508 | |||
| NM_007107.4 | NP_009038.1 | 5 | 619 | |||
| GenBank mRNA | AF087907.1 | - | 5 | 567 | ||
| AF110647.1 | - | 5 | 564 | |||
| AK304364.1 | - | 5 | 556 | |||
| AK312935.1 | - | 5 | 526 | |||
| BC017203.1 | - | 5 | 565 | 3618589 | ||
| BT006669.1 | - | 5 | 508 | |||
| DA283706.1 | - | 5 | 484 | |||
| DA725890.1 | - | 5 | 510 | |||

SSR3
Genomic Map