siRNA Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006948.4 | NP_008879.3 | 5 | 947 | ||
| GenBank mRNA | AK128540.1 | - | 5 | 729 | ||
| AK223282.1 | - | 5 | 905 | |||
| AK299004.1 | - | 4 | 682 | |||
| AK312396.1 | - | 5 | 902 | |||
| BC036370.2 | - | 5 | 909 | 4837136 | ||
| U04735.1 | - | 5 | 914 | |||

HSPA13
Genomic Map