siRNA Details
| Gene Symbol: STIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277961.1 | NP_001264890.1 | 8 | 1639 | ||
| NM_001277962.1 | NP_001264891.1 | 8 | 1639 | |||
| NM_003156.3 | NP_003147.2 | 8 | 1639 | |||
| GenBank mRNA | AK314928.1 | - | 8 | 1160 | ||
| BC021300.2 | - | 8 | 1631 | 4899542 | ||
| HQ317451.1 | - | 8 | 1638 | |||
| U52426.1 | - | 8 | 1636 | |||

STIM1
Genomic Map