siRNA Details
| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005642.2 | NP_005633.2 | 1 | 875 | ||
| GenBank mRNA | AK314837.1 | - | 1 | 1009 | ||
| BC022798.1 | - | 1 | 853 | |||
| BC032737.1 | - | 1 | 833 | 5539359 | ||
| U18062.1 | - | 1 | 811 | |||
| X97999.1 | - | 1 | 858 | |||

TAF7
Genomic Map