siRNA Details
| Gene Symbol: TALDO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006755.1 | NP_006746.1 | 4 | 469 | ||
| GenBank mRNA | AK295537.1 | - | 4 | 437 | ||
| AK313427.1 | - | 4 | 437 | |||
| BC010103.2 | - | 4 | 474 | 3536661 | ||
| BC018847.2 | - | 4 | 417 | |||
| L19437.2 | - | 4 | 469 | |||

TALDO1
Genomic Map