siRNA Details
| Gene Symbol: BUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278616.1 | NP_001265545.1 | 20 | 2598 | ||
| NM_001278617.1 | NP_001265546.1 | 21 | 2658 | |||
| NM_004336.4 | NP_004327.1 | 21 | 2658 | |||
| GenBank mRNA | AF011387.1 | - | 13 | 1723 | ||
| AF043294.2 | - | 21 | 2578 | |||
| AF046078.1 | - | 21 | 2595 | |||
| AF047471.1 | - | 21 | 2581 | |||
| AF053305.1 | - | 21 | 2609 | |||
| AK225317.1 | - | 21 | 2614 | |||
| AK291221.1 | - | 21 | 2611 | |||
| AK302418.1 | - | 20 | 2554 | |||
| BC028201.1 | - | 21 | 2579 | 5205385 | ||

BUB1
Genomic Map