siRNA Details
| Gene Symbol: TMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007114.2 | NP_009045.2 | 2 | 548 | ||
| XM_011534056.2 | XP_011532358.1 | 2 | 612 | |||
| GenBank mRNA | AB209109.1 | - | 2 | 518 | ||
| AK312697.1 | - | 2 | 502 | |||
| BC033771.2 | - | 2 | 493 | |||
| BC117418.1 | - | 2 | 401 | 40125969 | ||
| BC126123.1 | - | 2 | 401 | 8991839 | ||
| BC143840.1 | - | 2 | 401 | 9052355 | ||

TMF1
Genomic Map