siRNA Details
| Gene Symbol: LINC01137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038842.1 | 1 | 200 | |||
| GenBank mRNA | BM923085.1 | - | 1 | 146 | ||
| DA727774.1 | - | 1 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01137| Gene Symbol: LINC01137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038842.1 | 1 | 200 | |||
| GenBank mRNA | BM923085.1 | - | 1 | 146 | ||
| DA727774.1 | - | 1 | 203 | |||