siRNA Details
| Gene Symbol: BMPR1B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121610.1 | 2 | 183 | |||
| GenBank mRNA | AI150588.1 | - | 2 | 325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BMPR1B-AS1| Gene Symbol: BMPR1B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121610.1 | 2 | 183 | |||
| GenBank mRNA | AI150588.1 | - | 2 | 325 | ||