siRNA Details
| Gene Symbol: CCDC169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144981.2 | NP_001138453.1 | 4, 5 | 447 | ||
| NM_001144982.2 | NP_001138454.1 | 3, 4 | 367 | |||
| NM_001144983.2 | NP_001138455.1 | 3, 4 | 363 | |||
| NM_001144985.2 | NP_001138457.1 | 3, 4 | 367 | |||
| NM_001144986.2 | NP_001138458.1 | 3, 4 | 363 | |||
| NM_001198908.1 | NP_001185837.1 | 4, 5 | 447 | |||
| GenBank mRNA | AK308999.1 | - | 3, 4 | 336 | ||
| BC146975.1 | - | 4, 5 | 341 | 9056903 | ||
| BC171867.1 | - | 3, 4 | 258 | 9054521 | ||
| BC171872.1 | - | 3, 4 | 261 | 9054526 | ||
| BC171880.1 | - | 3, 4 | 257 | 9054534 | ||
| BC171882.1 | - | 3, 4 | 257 | 9054536 | ||
| BC171884.1 | - | 3, 4 | 261 | |||
| DN999189.1 | - | 4, 5 | 404 | |||

CCDC169
Genomic Map