siRNA Details
| Gene Symbol: SKP1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036619.1 | 1 | 789 | |||
| GenBank mRNA | AK310444.1 | - | 1 | 789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKP1P2| Gene Symbol: SKP1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036619.1 | 1 | 789 | |||
| GenBank mRNA | AK310444.1 | - | 1 | 789 | ||