siRNA Details
| Gene Symbol: OSBPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174060.1 | NP_001167531.1 | 1 | 40 | ||
| NM_017784.4 | NP_060254.2 | 1 | 40 | |||
| GenBank mRNA | AI798005.1 | - | 1 | 383 | ||
| Gene Symbol: ZNF860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137674.2 | NP_001131146.2 | 1 | 34 | ||
| GenBank mRNA | AK294003.1 | - | 1 | 34 | ||

OSBPL10
Genomic Map