siRNA Details
| Gene Symbol: LINC01346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040065.1 | 1, 2 | 78 | |||
| GenBank mRNA | AK124708.1 | - | 1, 2 | 73 | ||
| DB288420.1 | - | 1, 2 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01346| Gene Symbol: LINC01346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040065.1 | 1, 2 | 78 | |||
| GenBank mRNA | AK124708.1 | - | 1, 2 | 73 | ||
| DB288420.1 | - | 1, 2 | 125 | |||