siRNA Details
| Gene Symbol: LINC00977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033916.1 | 2, 3 | 222 | |||
| GenBank mRNA | BX090073.1 | - | 2, 3 | 222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00977| Gene Symbol: LINC00977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033916.1 | 2, 3 | 222 | |||
| GenBank mRNA | BX090073.1 | - | 2, 3 | 222 | ||