siRNA Details
Gene Symbol: LINC00977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033916.1 | 1, 2 | 71 | |||
GenBank mRNA | BX090073.1 | - | 1, 2 | 71 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033916.1 | 1, 2 | 71 | |||
GenBank mRNA | BX090073.1 | - | 1, 2 | 71 |