siRNA Details
| Gene Symbol: TULP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289395.1 | NP_001276324.1 | 9, 10 | 894 | ||
| NM_003322.4 | NP_003313.3 | 10, 11 | 1053 | |||
| GenBank mRNA | AB593123.1 | - | 9, 10 | 1050 | ||
| AB593124.1 | - | 10, 11 | 1039 | |||
| BC032714.1 | - | 9, 10 | 879 | 5495136 | ||
| BC065261.1 | - | 9, 10 | 862 | 6144003 | ||
| U82468.1 | - | 10, 11 | 1045 | |||

TULP1
Genomic Map