siRNA Details
| Gene Symbol: C1GALT1C1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101330.2 | NP_001094800.1 | 1 | 662 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1GALT1C1L| Gene Symbol: C1GALT1C1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101330.2 | NP_001094800.1 | 1 | 662 | ||