siRNA Details
| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170905.2 | NP_001164376.1 | 3 | 483 | ||
| NM_001294255.1 | NP_001281184.1 | 3 | 483 | |||
| GenBank mRNA | AK301806.1 | - | 3 | 327 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF736| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170905.2 | NP_001164376.1 | 3 | 483 | ||
| NM_001294255.1 | NP_001281184.1 | 3 | 483 | |||
| GenBank mRNA | AK301806.1 | - | 3 | 327 | ||