siRNA Details
| Gene Symbol: KCNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011544184.2 | XP_011542486.1 | 1 | 292 | ||
| GenBank mRNA | AK090902.1 | - | 1 | 292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK1| Gene Symbol: KCNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011544184.2 | XP_011542486.1 | 1 | 292 | ||
| GenBank mRNA | AK090902.1 | - | 1 | 292 | ||