siRNA Details
| Gene Symbol: CCDC61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267723.1 | NP_001254652.1 | 12, 13 | 1411 | ||
| XM_005259191.3 | XP_005259248.1 | 12, 13 | 3516 | |||
| XM_005259192.4 | XP_005259249.1 | 12, 13 | 2157 | |||
| XM_011527257.2 | XP_011525559.1 | 12, 13 | 1440 | |||
| GenBank mRNA | CA775840.1 | - | 4, 5 | 349 | ||
| CB852675.1 | - | 5, 6 | 398 | |||
| GQ375050.1 | - | 8, 9 | 844 | |||

CCDC61
Genomic Map