siRNA Details
| Gene Symbol: RAD51AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099218.2 | NP_001092688.1 | 1 | 812 | ||
| NM_001321233.1 | NP_001308162.1 | 5 | 1170 | |||
| XM_011533084.1 | XP_011531386.1 | 1 | 812 | |||
| GenBank mRNA | AK310498.1 | - | 6 | 1417 | ||
| DQ860102.1 | - | 1 | 812 | |||

RAD51AP2
Genomic Map