siRNA Details
| Gene Symbol: LINC01080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104138.1 | 2 | 340 | |||
| GenBank mRNA | DA225206.1 | - | 2 | 304 | ||
| DA346981.1 | - | 2 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01080| Gene Symbol: LINC01080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104138.1 | 2 | 340 | |||
| GenBank mRNA | DA225206.1 | - | 2 | 304 | ||
| DA346981.1 | - | 2 | 282 | |||