siRNA Details
| Gene Symbol: MMP24-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102705.1 | 1 | 25 | |||
| NR_102706.1 | 1 | 25 | ||||
| GenBank mRNA | HY005765.1 | - | 1 | 25 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMP24-AS1| Gene Symbol: MMP24-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102705.1 | 1 | 25 | |||
| NR_102706.1 | 1 | 25 | ||||
| GenBank mRNA | HY005765.1 | - | 1 | 25 | ||