siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 6 | 1067 | |||
| GenBank mRNA | DV080387.1 | - | 1 | 155 | ||
| DW009696.1 | - | 5 | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf173| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 6 | 1067 | |||
| GenBank mRNA | DV080387.1 | - | 1 | 155 | ||
| DW009696.1 | - | 5 | 550 | |||