siRNA Details
| Gene Symbol: CCDC175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164399.1 | NP_001157871.1 | 19 | 2347 | ||
| XM_011537120.2 | XP_011535422.1 | 19 | 2787 | |||
| XM_011537122.2 | XP_011535424.1 | 17 | 2564 | |||
| XM_011537124.2 | XP_011535426.1 | 16 | 2411 | |||

CCDC175
Genomic Map