siRNA Details
| Gene Symbol: CCDC175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164399.1 | NP_001157871.1 | 4 | 491 | ||
| XM_011537120.2 | XP_011535422.1 | 4 | 928 | |||
| XM_011537122.2 | XP_011535424.1 | 4 | 927 | |||
| XM_011537124.2 | XP_011535426.1 | 4 | 927 | |||
| XM_011537125.2 | XP_011535427.1 | 4 | 932 | |||
| XM_011537130.2 | XP_011535432.1 | 4 | 930 | |||
| XM_017021621.1 | XP_016877110.1 | 4 | 930 | |||

CCDC175
Genomic Map