siRNA Details
| Gene Symbol: CCDC175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164399.1 | NP_001157871.1 | 6, 7 | 905 | ||
| XM_011537120.2 | XP_011535422.1 | 6, 7 | 1342 | |||
| XM_011537122.2 | XP_011535424.1 | 6, 7 | 1341 | |||
| XM_011537124.2 | XP_011535426.1 | 6, 7 | 1341 | |||
| XM_011537125.2 | XP_011535427.1 | 6, 7 | 1346 | |||
| XM_011537130.2 | XP_011535432.1 | 6, 7 | 1344 | |||
| XM_017021621.1 | XP_016877110.1 | 6, 7 | 1344 | |||

CCDC175
Genomic Map