siRNA Details
| Gene Symbol: LOC146880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026899.1 | 3 | 832 | |||
| NR_027487.2 | 2 | 620 | ||||
| GenBank mRNA | AK055342.1 | - | 2 | 703 | ||
| BC032382.1 | - | 2 | 620 | |||
| BC120976.1 | - | 2 | 784 | |||
| BC120977.2 | - | 2 | 784 | |||

LOC146880
Genomic Map