siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109977.1 | NP_001103447.1 | 13, 14 | 3440 | ||
| XM_005263199.1 | XP_005263256.1 | 12, 13 | 3315 | |||
| XM_011532220.1 | XP_011530522.1 | 11, 12 | 3193 | |||
| XM_011532221.2 | XP_011530523.1 | 13, 14 | 3513 | |||
| XM_011532222.2 | XP_011530524.1 | 12, 13 | 3435 | |||
| XM_011532223.2 | XP_011530525.1 | 11, 12 | 3310 | |||
| XM_011532224.2 | XP_011530526.1 | 11, 12 | 3009 | |||
| XM_017008583.1 | XP_016864072.1 | 5, 6 | 1770 | |||
| GenBank mRNA | AK125363.1 | - | 9, 10 | 1673 | ||

FAM160A1
Genomic Map