siRNA Details
| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 2 | 611 | |||
| GenBank mRNA | AK054965.1 | - | 2 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01530| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 2 | 611 | |||
| GenBank mRNA | AK054965.1 | - | 2 | 608 | ||