siRNA Details
| Gene Symbol: FAM166B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099951.3 | NP_001093421.1 | 3 | 385 | ||
| NM_001164310.2 | NP_001157782.1 | 3 | 385 | |||
| NM_001287238.1 | NP_001274167.1 | 3 | 385 | |||
| NM_001287239.1 | NP_001274168.1 | 3 | 385 | |||
| XM_011518028.2 | XP_011516330.1 | 3 | 524 | |||
| GenBank mRNA | BC129999.1 | - | 3 | 385 | 40127800 | |
| BC146928.1 | - | 3 | 385 | 9056856 | ||
| BC146933.1 | - | 3 | 385 | 9056861 | ||
| BC171840.1 | - | 3 | 385 | 9054494 | ||
| BC171851.1 | - | 3 | 385 | 9054505 | ||

FAM166B
Genomic Map