siRNA Details
| Gene Symbol: TMEM179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286389.1 | NP_001273318.1 | 4 | 2361 | ||
| XM_011536745.2 | XP_011535047.1 | 5 | 3088 | |||
| XM_011536750.2 | XP_011535052.1 | 5 | 2273 | |||
| GenBank mRNA | AF462446.1 | - | 1 | 671 | ||
| BC047917.1 | - | 1 | 318 | |||

TMEM179
Genomic Map