siRNA Details
Gene Symbol: TYRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006293.3 | NP_006284.2 | 17 | 2312 | ||
XM_017022542.1 | XP_016878031.1 | 17 | 2277 | |||
XM_017022543.1 | XP_016878032.1 | 17 | 2113 | |||
GenBank mRNA | AB209431.1 | - | 17 | 2341 | ||
AK295496.1 | - | 6 | 607 | |||
BC029925.2 | - | 9 | 962 | |||
BC034960.1 | - | 17 | 2094 | |||
BC049368.1 | - | 17 | 2086 | 4821587 | ||
BC051756.1 | - | 17 | 2091 | 6095596 | ||
D17517.1 | - | 17 | 2300 | |||
D50479.1 | - | 17 | 2072 | |||
U02566.1 | - | 14 | 1520 | |||
U05682.1 | - | 17 | 2082 | |||
X72886.1 | - | 6 | 522 |
