siRNA Details
| Gene Symbol: LOC105374250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322103.1 | NP_001309032.1 | 3 | 348 | ||
| XM_011513348.2 | XP_011511650.1 | 2 | 167 | |||
| XM_011513349.2 | XP_011511651.1 | 2 | 163 | |||
| XM_011513350.2 | XP_011511652.1 | 2 | 162 | |||
| GenBank mRNA | BX116842.1 | - | 2 | 75 | ||
| DB090841.1 | - | 3 | 348 | |||
| HY027726.1 | - | 2 | 120 | |||

LOC105374250
Genomic Map