siRNA Details
Gene Symbol: CSN1S2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033311.1 | 3, 4 | 120 | |||
GenBank mRNA | AW104440.1 | - | 4, 5 | 320 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CSN1S2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033311.1 | 3, 4 | 120 | |||
GenBank mRNA | AW104440.1 | - | 4, 5 | 320 |