siRNA Details
Gene Symbol: CSN1S2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033311.1 | 6, 7 | 306 | |||
GenBank mRNA | AW104440.1 | - | 2, 3 | 134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CSN1S2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033311.1 | 6, 7 | 306 | |||
GenBank mRNA | AW104440.1 | - | 2, 3 | 134 |