siRNA Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000587.2 | NP_000578.2 | 13 | 1807 | ||
| GenBank mRNA | AK290349.1 | - | 13 | 1817 | ||
| AK304848.1 | - | 12 | 1280 | |||
| AK312737.1 | - | 13 | 1741 | |||
| BC063851.1 | - | 13 | 1788 | 6184213 | ||
| J03507.1 | - | 12 | 1693 | |||

C7
Genomic Map