siRNA Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000587.2 | NP_000578.2 | 14 | 1915 | ||
| GenBank mRNA | AK290349.1 | - | 14 | 1925 | ||
| AK304848.1 | - | 13 | 1388 | |||
| AK312737.1 | - | 14 | 1849 | |||
| BC063851.1 | - | 14 | 1896 | 6184213 | ||
| J03507.1 | - | 13 | 1801 | |||

C7
Genomic Map