siRNA Details
| Gene Symbol: LINC00202-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026794.1 | 1, 2 | 5186 | |||
| GenBank mRNA | CR749234.1 | - | 1, 2 | 5126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00202-2| Gene Symbol: LINC00202-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026794.1 | 1, 2 | 5186 | |||
| GenBank mRNA | CR749234.1 | - | 1, 2 | 5126 | ||