siRNA Details
| Gene Symbol: UBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003334.3 | NP_003325.2 | 16 | 2038 | ||
| NM_153280.2 | NP_695012.1 | 16 | 1966 | |||
| XM_005272649.1 | XP_005272706.1 | 17 | 2416 | |||
| XM_011543954.2 | XP_011542256.1 | 17 | 2058 | |||
| XM_017029777.1 | XP_016885266.1 | 16 | 3133 | |||
| XM_017029778.1 | XP_016885267.1 | 17 | 4307 | |||
| XM_017029779.1 | XP_016885268.1 | 16 | 4815 | |||
| XM_017029780.1 | XP_016885269.1 | 16 | 1923 | |||
| XM_017029781.1 | XP_016885270.1 | 16 | 1971 | |||
| GenBank mRNA | AK293161.1 | - | 5 | 630 | ||
| AK296866.1 | - | 6 | 656 | |||
| BC013041.2 | - | 16 | 1905 | 3543021 | ||
| BC020261.1 | - | 13 | 1600 | |||
| M58028.1 | - | 16 | 1944 | |||
| X52897.1 | - | 9 | 1110 | |||
| X56976.1 | - | 16 | 1847 | |||

UBA1
Genomic Map