siRNA Details
| Gene Symbol: TEX22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195082.1 | NP_001182011.1 | 4 | 1262 | ||
| XM_006720234.3 | XP_006720297.1 | 4 | 1536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX22| Gene Symbol: TEX22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195082.1 | NP_001182011.1 | 4 | 1262 | ||
| XM_006720234.3 | XP_006720297.1 | 4 | 1536 | |||