siRNA Details
| Gene Symbol: MYRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127392.2 | NP_001120864.1 | 10 | 1565 | ||
| NM_013279.3 | NP_037411.1 | 10 | 1465 | |||
| XM_005274222.1 | XP_005274279.1 | 10 | 1553 | |||
| XM_005274223.1 | XP_005274280.1 | 10 | 1553 | |||
| XM_005274224.1 | XP_005274281.1 | 10 | 1553 | |||
| XM_005274225.1 | XP_005274282.1 | 10 | 1553 | |||
| XM_005274226.1 | XP_005274283.1 | 10 | 1553 | |||
| XM_005274227.1 | XP_005274284.1 | 10 | 1553 | |||
| XM_005274228.1 | XP_005274285.1 | 10 | 1400 | |||
| XM_011545234.2 | XP_011543536.1 | 10 | 1439 | |||
| GenBank mRNA | AB023171.2 | - | 10 | 1553 | ||
| AF086762.1 | - | 10 | 1465 | |||

MYRF
Genomic Map