siRNA Details
| Gene Symbol: WRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000553.4 | NP_000544.2 | 5 | 1223 | ||
| XM_011544639.2 | XP_011542941.1 | 5 | 705 | |||
| GenBank mRNA | AF091214.1 | - | 5 | 666 | ||
| AY818673.1 | - | 4 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WRN| Gene Symbol: WRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000553.4 | NP_000544.2 | 5 | 1223 | ||
| XM_011544639.2 | XP_011542941.1 | 5 | 705 | |||
| GenBank mRNA | AF091214.1 | - | 5 | 666 | ||
| AY818673.1 | - | 4 | 435 | |||